Learn More
Tight Junction Protein 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Marca:  Novus Biologicals NBP1-85046
 Ver más versiones de este producto
Código de producto. 18746573
  • ONLINE EXCLUSIVE PRICE
    572.00€
    540.75€ / 0.10 ml
    Ahorro 31.25€
    5% Descuento
Descripción
Tight Junction Protein 1 Polyclonal specifically detects Tight Junction Protein 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Polyclonal
IgG
Certificados
Promociones
Promociones
For Research Use Only