missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIA1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Descripción
The protein encoded by this gene is a member of a family of RNA-binding proteins, has three RNA recognition motifs , and binds adenine and uridine-rich elements in mRNA and pre-mRNAs of a wide range of genes. It regulates various activities including translational control, splicing and apoptosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. The different isoforms have been show to function differently with respect to post-transcriptional silencing.
Especificaciones
Especificaciones
| Antígeno | TIA1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2B1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | TIA1 |
| N.º de referencia del gen | P31483 |
| Alias de gen | TIA-1, WDM |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?