missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Antibody (2H11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-33070AF647
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| Thyroglobulin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| monoclonal antibody 2H11 reacts with a partially defined epitope of human thyroglobulin. This epitope is different form the epitope recognized by monoclonal antibody 6E1. Thyroglobulin is a 660kDa dimeric pre-protein with multiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 2H11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
| TG | |
| Human thyroid follicular cells were used as immunogen to generate the antibody. | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 7038 | |
| Human, Mouse, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido