missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thrombomodulin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA586697
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Purity is > 95% by SDS-PAGE.
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
Especificaciones
| Thrombomodulin | |
| Polyclonal | |
| Unconjugated | |
| THBD | |
| AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21824, 7056, 83580 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07204, P15306 | |
| THBD | |
| Synthetic peptide, corresponding to amino acids 530-575 of Human Thrombomodulin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido