Learn More
DiscoverX THIQ
Código de producto. 17382589
Encompass
Click to view available options
:
1 mg
Código de producto. 17382589
Marca: DiscoverX 921278
THIQ is an agonist for MC4R. Provided as 1mg of dry powder.
THIQ is a synthetic drug molecule and selective agonist for the melanocortin receptor MC4 over MC3 and MC1. Stimulates sexual activity, including enhancement in intracavernosal pressure, but has little effect on appetite or inflammation. Provided as 1mg of dry powder.
Especificaciones
Cantidad 1 mg