Learn More
Thiamphenicol, MedChemExpress
Código de producto. 16412156
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 mg
500 mg
Tamaño de la unidad:
100 miligramos
500 miligramos
16412156 500 mg 500 miligramos
16428336 100 mg 100 miligramos
2 options
Código de producto. 16412156
Marca: Medchem Express HYB0479
MedChemExpress Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).
Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).
Especificaciones
Nombre del producto químico o material Thiamphenicol
Forma física Solid
CAS 15318-45-3
Color Blanco
Cantidad 500 mg
Notas de grado de pureza Research
Fórmula molecular C12H15Cl2NO5S
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo Thiophenicol Dextrosulphenidol
Información de solubilidad DMSO : 100 mg/mL (280.73 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.61 mM; Need ultrasonic)
SMILES O=C(N[C@H](CO)[C@H](O)C1=CC=C(S(=O)(C)=O)C=C1)C(Cl)Cl
Peso molecular (g/mol) 356.22
Formula Weight (peso de la fórmula) 356.22
Porcentaje de pureza 99.38%