Learn More
Cole-Parmer Thermoplastic Vulcanizate (TPV) Pharma Grade Tubing
Código de producto. 30054171 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/8 in
3/16 in
3/8 in
Diámetro exterior (imperial):
1/2 in
1/4 in
3/16 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
Tamaño de la unidad:
15 metros
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30054171 1/16 in 3/16 in 15 metros
30053527 1/2 in 3/4 in 15 metros
30053912 1/4 in 1/2 in 15 metros
30054338 3/16 in 7/16 in 15 metros
30053517 1/4 in 7/16 in 15 metros
30053451 1/4 in 3/8 in 15 metros
30053658 3/16 in 5/16 in 15 metros
30054210 1/8 in 1/4 in 15 metros
30053991 3/8 in 5/8 in 15 metros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Código de producto. 30054171 Proveedor Cole-Parmer N.º de proveedor 5011982
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Offers great chemical and kink resistance for e in., cellent pump performance
Biocompatible thermoplastic tubing offers good chemical resistance. Does not contain BPA, phthalates, PVC, or pero in., ides. Sterilize by autoclave, gamma irradiation, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Certificaciones/conformidad FDA;Non-Phthalate;USP;RoHS
Diámetro interior (imperial) 1/16 in
Diámetro exterior (imperial) 3/16 in
Longitud global (imperial) 50 ft.