Learn More
- Home
- Products
- Tubos
- Tubos de silicona y caucho
- Cole-Parmer Thermoplastic Elastomer (TPE) Tubing
Cole-Parmer Thermoplastic Elastomer (TPE) Tubing
Código de producto. p-200105032 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/32 in
1/4 in
1/8 in
1 in
3/16 in
3/32 in
3/4 in
3/8 in
5/16 in
5/8 in
Diámetro exterior (imperial):
1-1/4 in
1/2 in
1/4 in
1/8 in
1 in
3/16 in
3/32 in
3/4 in
3/8 in
5/16 in
5/32 pulg
5/8 in
7/16 in
7/32 pulg
7/8 in
9/16 in
Tamaño de la unidad:
15 metros
| Código de producto. | Diámetro interior (imperial) | Diámetro exterior (imperial) | unitSize |
|---|---|---|---|
| 30053953 | 5/16 in | 7/16 in | 15 metros |
| 30054322 | 1/2 in | 3/4 in | 15 metros |
| 30053061 | 1/16 in | 1/8 in | 15 metros |
| 30054035 | 3/32 in | 7/32 pulg | 15 metros |
| 30053555 | 3/4 in | 1 in | 15 metros |
| 30054199 | 1/8 in | 3/16 in | 15 metros |
| 30053882 | 1/4 in | 3/8 in | 15 metros |
| 30053623 | 3/32 in | 5/32 pulg | 15 metros |
| 30053690 | 3/8 in | 1/2 in | 15 metros |
| 30053210 | 1/8 in | 1/4 in | 15 metros |
| 30053893 | 1/4 in | 7/16 in | 15 metros |
| 30054004 | 3/8 in | 5/8 in | 15 metros |
| 30053446 | 1/32 in | 3/32 in | 15 metros |
| 30053382 | 3/16 in | 5/16 in | 15 metros |
| 30053920 | 3/16 in | 3/8 in | 15 metros |
| 30053435 | 1/16 in | 3/16 in | 15 metros |
| 30053579 | 5/8 in | 7/8 in | 15 metros |
| 30054113 | 3/16 in | 7/16 in | 15 metros |
| 30054236 | 5/16 in | 1/2 in | 15 metros |
| 30054138 | 1/4 in | 1/2 in | 15 metros |
| 30053316 | 3/8 in | 9/16 in | 15 metros |
| 30053060 | 3/16 in | 1/4 in | 15 metros |
| 30054010 | 3/4 in | 7/8 in | 15 metros |
| 30053198 | 1 in | 1-1/4 in | 15 metros |
Este artículo no se puede devolver.
Vea la política de devoluciones
Código de producto. 30053953
Marca: Cole-Parmer 5011613
Inicie sesión o regístrese para comprobar su precio y disponibilidad.
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diámetro interior (imperial) | 5/16 in |
| Material | TPE |
| Diámetro exterior (imperial) | 7/16 in |
| Longitud global (imperial) | 50 ft. |
| Longitud global (métrico) | 50 ft. |
| Método de esterilización | E-beam/gamma irradiation between 25 and 45 kGy |
| Color | Clear |