Learn More
Thermo Scientific™ Aegis™ 5-14 Single-Use BioProcess Container (BPC), with 2 ports, 2D
GREENER_CHOICE
Código de producto. 17835716
Change view
Click to view available options
Capacidad (métrico):
50 mL
100 mL
250 mL
500 mL
1 L
Tamaño de la unidad:
paquete de 10
Código de producto. Capacidad (métrico) unitSize
17835716 100 mL paquete de 10
17895706 50 mL paquete de 10
17845716 250 mL paquete de 10
17865716 500 mL paquete de 10
17875716 1 L paquete de 10
5 options
Código de producto. 17835716
Marca: Thermo Scientific™ SH30961.12
Thermo Scientific™ Labtainer BPCs are composed of Thermo Scientific Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Thermo Scientific™ Labtainer BPCs are composed of Thermo Scientific Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
• All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Flexible and economic solution to update or increase your bioreactor capacity
• Suitable for both suspension and adherent cultures
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and gas filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Specifications
Capacidad (métrico) 100 mL
Dimensiones 14.7 x 14.2 cm (5.8 x 5.6 in.)
Detalle del puerto Two Edge Ports, Female and Male Luer Lock Connections
Cantidad 10/Cs.
Método de esterilización Gamma Irradiated
Línea de longitud de tuberías 1 30 cm (12 in.)
Línea de longitud de tuberías 2 30 cm (12 in.)
Línea de material de tuberías 1 C-flex
Línea de material de tuberías 2 C-flex