Learn More
Thermo Scientific™ 48-Well Semi-Skirted Plates
Código de producto. 12687416
Change view
Click to view available options
Color:
Blanco
Morado
Verde
Tamaño de la unidad:
paquete de 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Color unitSize
12687416 Blanco paquete de 50
10640201 Morado paquete de 50
10140653 Verde paquete de 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 12687416 Proveedor Thermo Scientific™ N.º de proveedor AB0648/W
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
48-well semi-skirted plate for PCR and qPCR applications.
The Thermo Scientific 48-well semi-skirted plate for PCR and qPCR applications comes in a convenient format, which allows for lower sample numbers. The semi skirt makes the plate more rigid and allots space for labeling and barcoding. Available in assorted colors, it's compatible with standard multichannel pipettes and automated liquid handling systems.The plate is sealable when used with adhesive films, foils or cap strips.
Highlights
• Convenient format for lower sample numbers
• Semi-skirt adds rigidity and allows for labeling or barcoding
• Sealable using adhesive films and foils and flat, ultra clear and domed cap strips
• 0.3 mL maximum well volume (when used with adhesive and heat seals)
TRUSTED_SUSTAINABILITY
Especificaciones
Formato Estándar
matriz, red, conjunto 6 x 8
Color Blanco
Material Virgin Polypropylene
N.° de pocillos 48