Learn More
TGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16870864 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16870864 20 μL 20 microlitros
16860864 150 μL 150 microlitros
2 options
Código de producto. 16870864
Marca: Proteintech 129123AP20UL
Rabbit Polyclonal Antibody
This antibody recognizes the endogenous 90 kDa TGM1 proteins.
The protein encoded by this gene is a membrane protein that catalyzes the addition of an alkyl group from an akylamine to a glutamine residue of a protein, forming an alkylglutamine in the protein. This protein alkylation leads to crosslinking of proteins and catenation of polyamines to proteins. This gene contains either one or two copies of a 22 nt repeat unit in its 3′ UTR. Mutations in this gene have been associated with autosomal recessive lamellar ichthyosis (LI) and nonbullous congenital ichthyosiform erythroderma (NCIE).
Especificaciones
Antígeno TGM1
Aplicaciones Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica TGM1
N.º de referencia del gen P22735, P23606, Q9JLF6
Alias de gen Epidermal TGase, ICR2, KTG, LI, LI1, TG(K), TGASE, TGase 1, TGase K, TGK, TGM1, Transglutaminase 1, Transglutaminase K
Símbolos de los genes Tgm1
Especie del huésped Rabbit
Inmunógeno TGM1 Fusion Protein Ag3905
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21816, 60335, 7051
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG