missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGF beta-1 Monoclonal Antibody (TB21)
GREENER_CHOICE

Código de producto. 15394291
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15394291 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15394291

Marca: Invitrogen™ MA518023

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This antibody reacts with the dimeric (25 kDa) and monomeric (12.5 kDa) forms of TGF-beta under both non-reducing and reducing conditions. In ELISA applications, this antibody recognizes both human platelet-derived and recombinant TGF-beta. This antibody neutralizes TGF-beta activity in vitro in an inhibition assay of CCL/64 cell growth, and neutralizes the growth promoting action of TGF-beta in the NRK-49F colony forming assay. The effect of microinjection of this antibody into one blastomere of two cell stage Xenopus embryos indicated that it was able to neutralize effectively the bioactivity of TGF-beta in vivo. In IHC applications, this antibody has been used to demonstrate TGF-beta in ovine ovarian tissue and human breast carcinoma. As a consequence of the intense staining of the erythrocytes, it is possible to locate a single cell within the ovarian stroma making it useful in locating very fine capillary networks within tissue.

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TGF beta-1
Aplicaciones ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon TB21
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7
génica Tgfb1
N.º de referencia del gen P01137, P04202, P17246, P18341, P50414
Alias de gen CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product
Símbolos de los genes Tgfb1
Especie del huésped Mouse
Inmunógeno TGF-beta from human platelets.
Método de purificación Protein G
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100008645, 21803, 282089, 443417, 59086, 7040
Especies diana Human, Mouse, Rat, Rabbit, Bovine, Sheep, Mink
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.