Learn More
Invitrogen™ TGF beta-1,2,3 Monoclonal Antibody (1D11)
GREENER_CHOICE
Código de producto. 15583482
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
15583482 500 μg 500 microgramos
1 options
Código de producto. 15583482 Proveedor Invitrogen™ N.º de proveedor MA523795
Mouse Monoclonal Antibody
This antibody recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method. Neutralization: measured by its ability to neutralize TGF beta-1 inhibition of IL-4-dependent proliferation in the HT-2 mouse T cell line. The Neutralization Dose (ND50) is typically 0.25-1.25 μg/mL in the presence of 1 ng/mL Recombinant Human TGF beta-1 and 7.5 ng/mL Recombinant Mouse IL-4.
TGF-beta1, -2, and -3 are a closely related group of proteins (70-80% sequence homology) that are produced by many cell types and function as growth and differentiation factors. The active forms of TGF-beta1, -2, and -3 are disulfide-linked homodimers.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TGF beta-1,2,3
Aplicaciones ELISA, Inhibition Assays, Neutralization, Western Blot
Clasificación Monoclonal
Clon 1D11
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and No Preservative; pH 7.4
génica TGFB1
N.º de referencia del gen P01137, P04202, P09531, P10600, P16047, P17125, P18341, P21214, P27090, P30371, P61812
Alias de gen CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; tgf beta 1; tgf beta 3; tgf beta1; TGFB; Tgfb1; Tgfb-1; TGFB4; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; TGF-beta4; tissue growth factor-beta4; Transforming growth factor; transforming growth factor beta 1; transforming growth factor beta 4; transforming growth factor beta 4 precursor; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta-1 precursor
Símbolos de los genes TGFB1, TGFB2, TGFB3
Especie del huésped Mouse
Inmunógeno Bovine bone-derived TGF-beta 1 and TGF-beta 2.
Método de purificación Protein A/G
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100873157, 21803, 21808, 21809, 282089, 396438, 421352, 534069, 538957, 7040, 7042, 7043
Especies diana Human, Mouse, Bovine, Chicken
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1