Learn More
TGF-beta 1, 2, 3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB1835R-01M
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Neutralization.
Especificaciones
| TGF beta 1/2/3 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7040 | |
| Multi-species | |
| IgG1 |
| Neutralization | |
| 1D11R | |
| Neutralization 0.25-1.25 ug/mL | |
| ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1 | |
| Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
| 1 mg | |
| Primary | |
| Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Título del producto
Spot an opportunity for improvement?Share a Content Correction