missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TER-119 Monoclonal Antibody (TER119), PE
GREENER_CHOICE

Código de producto. 15324101
Change view
Click to view available options
Cantidad:
300 μg
50 μg
Unit Size:
300 microgramos
50 microgramos
Product Code. Cantidad unitSize
15324101 50 μg 50 microgramos
15334101 300 μg 300 microgramos
2 options
This item is not returnable. View return policy

Product Code. 15324101

Brand: Invitrogen™ MA517823

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody

TER-119 Monoclonal Antibody for Flow

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TER-119
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon TER119
Concentración 0.1 mg/mL
Conjugado PE
Formulación PBS with 4-5mg/mL BSA and 0.02% sodium azide
génica Ly76
N.º de referencia del gen 0
Alias de gen Ly76; Ly-76; TER119
Símbolos de los genes Ly76
Especie del huésped Rat
Inmunógeno Day 14 BALB/c fetal liver cells.
Método de purificación Protein G
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 104231
Especies diana Mouse
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.