Learn More
TER-119/Erythroid Cells Rat anti-Mouse, PE, Clone: TER-119, BD
Descripción
The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage cocktail” used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Fetal Liver
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | TER-119/Erythroid Cells |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | TER-119 |
| Concentración | 0.2mg/mL |
| Conjugado | PE |
| Descripción | Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Mouse Fetal Liver |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.