missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TENR Polyclonal Antibody

Descripción
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (88%), Rat (86%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plays a role in spermatogenesis. Binds to RNA but not to DNA.
Especificaciones
Especificaciones
| Antígeno | TENR |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | ADAD1 |
| N.º de referencia del gen | Q96M93 |
| Alias de gen | ADAD1; adenosine deaminase domain containing 1; adenosine deaminase domain containing 1 (testis specific); adenosine deaminase domain containing 1 (testis-specific); adenosine deaminase domain-containing protein 1; TENR; testis nuclear RNA binding protein; testis nuclear RNA-binding protein; testis tissue sperm-binding protein Li 92mP |
| Símbolos de los genes | ADAD1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?