missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEF1 Recombinant Rabbit Monoclonal Antibody (2E7N10)
GREENER_CHOICE

Código de producto. 17186333
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17186333 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17186333 Proveedor Invitrogen™ N.º de proveedor MA535679

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

TEF1 Recombinant Monoclonal Antibody for ChIP, CUT&RUN, ELISA, ICC/IF, IP, Western Blot

TEF1 (Transcriptional enhancer factor 1), a member of the TEA/ATTS domain family, is a nuclear protein that is expressed in numerous cell types and plays a role in controlling the expression of numerous genes. TEF family members have a highly conserved DNA-binding domain TEF-1 binds to GT-IIC, SphI/II and M-CAT. TEF-1 also binds to the proximal regulatory element (PRE) of transforming growth factor-alpha, a member of the EGF family that is overexpressed in many types of cancer. Furthermore, TEF-1 represses transcription in placental cells. In vitro, TEF-1 is phosphorylated by several PKC isozymes. TEF-1 is phosphorylated in vivo at serine and threonine residues. Phosphorylation of TEF-1, both in vivo and in vitro, results in a reduction in its DNA-binding capability, which suggests a potential role for TEF-1 in PKC inhibition. TEF-1 also complexes with larger tumor antigen (TAg), and may thus have a role in tumorigenesis. Dimerization of TEF-1 may be important for TEF-1 to function as a regulator of gene transcription.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TEF1
Aplicaciones ChIP Assay, ELISA, Immunoprecipitation, Western Blot, CUT&RUN, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 2E7N10
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
génica Tead1
N.º de referencia del gen P28347, P30051
Alias de gen 2610024B07Rik; AA; B230114H05Rik; Gtrgeo5; mTEF-1; N TEAD1; NTEF1; NTEF-1; protein GT-IIC; REF1; Tcf13; TCF-13; TEA domain family member 1; TEA domain family member 1 (SV40 transcriptional enhancer factor); TEA domain transcription factor 1; TEAD1; TEAD-1; TEF1; TEF-1; Transcription factor 13; transcriptional enhancer factor 1; transcriptional enhancer factor TEF-1
Símbolos de los genes Tead1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21676, 361630, 7003
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.