Learn More
TEAD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12418-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.Especificaciones
| TEAD4 | |
| Polyclonal | |
| Unconjugated | |
| TEAD4 | |
| EFTR 2, hRTEF 1B, RTEF1, TCF13L1, TEA domain family member 4, TEAD 4, TEAD4, TEF 3, TEF3, TEFR 1, Transcription factor 13 like 1, Transcription factor RTEF 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21679, 683522, 7004 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15561, Q62296 | |
| TEAD4 | |
| TEAD4 Fusion Protein Ag3059 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |