Learn More
TEAD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16860124 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16860124 150 μL 150 microlitros
16870124 20 μL 20 microlitros
2 options
Código de producto. 16860124
Marca: Proteintech 124181AP150UL
Rabbit Polyclonal Antibody
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
Especificaciones
Antígeno TEAD4
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica TEAD4
N.º de referencia del gen Q15561, Q62296
Alias de gen EFTR 2, hRTEF 1B, RTEF1, TCF13L1, TEA domain family member 4, TEAD 4, TEAD4, TEF 3, TEF3, TEFR 1, Transcription factor 13 like 1, Transcription factor RTEF 1
Símbolos de los genes TEAD4
Especie del huésped Rabbit
Inmunógeno TEAD4 Fusion Protein Ag3059
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21679, 683522, 7004
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG