Learn More
R&D Systems™ TdT In Situ Apoptosis Detection Kit - Replenisher
Descripción
Includes
Proteinase K Solution, 30uL; TdT dNTP Mix, 30uL; TdT Enzyme, 30uL; Strep-HRP, 30uL; 30uL each of 50X cations
The replenisher is also useful when using alternative detection and counterstaining methods in double-labeling experiments
Abbreviated instructions compared to the complete TACS 2 kits
Especificaciones
Especificaciones
| Cantidad | 30 pruebas |
| Contenido y almacenamiento | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |