missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TDO2 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15913535
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15913535 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15913535

Marca: Invitrogen™ PA577018

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Tryptophan 2,3-dioxygenase (TDO2) is an enzyme encoded by the TDO2 gene, predominantly expressed in the liver and playing a crucial role in tryptophan metabolism through the kynurenine pathway. TDO2 catalyzes the first and rate-limiting step of this pathway, converting tryptophan into N-formylkynurenine. This metabolic process is vital for maintaining systemic tryptophan homeostasis and regulating its availability for protein synthesis and serotonin production. Beyond its metabolic function, TDO2 is involved in modulating immune responses. It contributes to immune tolerance by depleting tryptophan, thereby inhibiting T cell proliferation and promoting regulatory T cell development. Recent research has identified TDO2's role in cancer, particularly in shaping the tumor microenvironment and facilitating immune escape mechanisms through tryptophan catabolism. Its expression is upregulated in several cancers, including gliomas, correlating with tumor severity and patient prognosis. TDO2 and its associated pathways have become therapeutic targets in cancer immunotherapy due to their influence on immune system modulation and metabolism.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TDO2
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
génica TDO2
N.º de referencia del gen P48775, P48776
Alias de gen AA407491; chky; HAMAP-Rule:MF_03020}; TDO; TDO {ECO:0000255; tdo variant1; tdo variant2; TDO2; TO; TO {ECO:0000255; TPH2; TRPO; TRPO {ECO:0000255; tryptamin 2,3-dioxygenase; tryptamin 2,3-dioxygenase {ECO:0000255; Tryptophan 2,3-dioxygenase; tryptophan 2,3-dioxygenase {ECO:0000255; tryptophan 2,3-dioxygenase A; tryptophan 2,3-dioxygenase A; tryptophan 2,3-dioxygenase; tryptophan oxygenase; tryptophan oxygenase {ECO:0000255; Tryptophan pyrrolase; tryptophan pyrrolase {ECO:0000255; tryptophan-2,3-dioxygenase; tryptophan-23-dioxygenase; tryptophanase; tryptophanase {ECO:0000255
Símbolos de los genes TDO2
Especie del huésped Rabbit
Inmunógeno Recombinant full length Human TDO2.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 56720, 6999
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.