missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 2 Monoclonal Antibody (B20.1), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Código de producto. 17850971
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17850971 100 μg 100 microgramos
17800971 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17850971 Proveedor Invitrogen N.º de proveedor M033T03Y03

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TCR V alpha 2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon B20.1
Concentración 0.1 mg/mL
Conjugado NovaFluor Yellow 610
Formulación PBS with 0.09% sodium azide; pH 7.2
Alias de gen TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2
Especie del huésped Rat
Método de purificación Affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento 4° C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a λ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.