missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR gamma/delta Monoclonal Antibody (V65)
GREENER_CHOICE

Código de producto. 11504423
Change view
Click to view available options
Cantidad:
250 μg
Tamaño de la unidad:
250 microgramos
Código de producto. Cantidad unitSize
11504423 250 μg 250 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11504423

Marca: Invitrogen™ MA170028

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA1-70028 detects gamma/delta TCRV65 in rat samples. MA1-70028 has been successfully used in flow cytometry, immunoprecipitation, immunohistochemistry (frozen sections) procedures. The MA1-70028 immunogen is rat T Blasts.

TCR gamma/delta (T-cell receptor gamma/delta) are specialized T-cells in the immune system. The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory auto-destructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). However, a small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TCR gamma/delta
Aplicaciones Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Clasificación Monoclonal
Clon V65
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
génica Trg
N.º de referencia del gen 0
Alias de gen RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@
Símbolos de los genes Trg
Especie del huésped Mouse
Inmunógeno Rat T Blasts.
Método de purificación Protein G
Cantidad 250 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 24821
Especies diana Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.