missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR beta Monoclonal Antibody (H57-597), NovaFluor™ Violet 800, eBioscience™

Código de producto. p-200089174
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30245297 25 μg 25 microgramos
30245412 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30245297 Proveedor Invitrogen™ N.º de proveedor M013T02V03A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Armenian Hamster Monoclonal Antibody

Description: The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested: The H57-597 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.6 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest Master mixes.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TCR beta
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon H57-597
Concentración 0.1 mg/mL
Conjugado NovaFluor Violet 800
Formulación PBS with BSA and 0.09% sodium azide; pH 7.2
génica Tcrb
Alias de gen bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA
Símbolos de los genes Tcrb
Especie del huésped Armenian Hamster
Método de purificación Affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21577
Especies diana Mouse
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.