missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA523385
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TCF19 Polyclonal Antibody for IHC (P)
TCF-19 (transcription factor 19), also known as SC1 or SC1-1, is a 345 amino acid protein that contains one FHA domain, a proline-rich domain and one PHD-type zinc finger. Localizing to the nucleus, TCF-19 is a growth regulated protein that is believed to function as a trans-activating factor with a role in the transcription of genes involved in the late stages of cell cycle progression (G1 to S transition or entry of cells into G2 and mitosis). TCF-19 is expressed preferentially in the G1-S phase of the cell cycle. The gene encoding TCF-19 localizes to a critical region on chromosome 6 that has been associated with psoriasis vulgaris, a disorder of the skin that is characterized by hyperproliferation of epidermal cells. This suggests that TCF-19, via its regulation of late cell cycle-specific genes, may play a role in the development of this disorder.
Especificaciones
| TCF19 | |
| Polyclonal | |
| Unconjugated | |
| TCF19 | |
| 5730403J10Rik; AW495861; DAMA-213L4.5; SC 1; SC1; SC1 protein homolog; SC1(TCF19)-6; SC1(TCF19)-7; SC1-1; SCI(TCF19)-4; TCF 19; TCF19; TCF-19; Transcription factor 19; transcription factor 19-like protein; Transcription factor SC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6941 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q9Y242 | |
| TCF19 | |
| A synthetic peptide corresponding to amino acids 259-287 of human TCF19. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?