missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Bile X-Glucuronide (TBX) Medium (Dehydrated)
GREENER_CHOICE

Código de producto. 13225719
Change view
Click to view available options
Cantidad:
500 g
5 kg
Tamaño de la unidad:
5 Kg
500 GR
Código de producto. Cantidad unitSize
13225719 5 kg 5 Kg
10622065 500 g 500 GR
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 13225719

Marca: Thermo Scientific™ CM0945T

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Detect and enumerate Escherichia coli in food and animal feed samples with this chromogenic medium while adhering to ISO 11133:2014.

Detect and enumerate Escherichia coli in food and animal feed samples with Thermo Scientific™ Oxoid™ Typtone Bile X-Glucuronide (TBX) Medium (Dehydrated). The medium meets the formulation and performance criterion laid down in ISO 166491, 2, 3 and ISO 11133:20144.

TBX Medium builds on the speed, reliability and recovery from frozen samples and detection of poor lactose fermenters offered by the original Tryptone Bile Agar4,5, with the addition of a chromogenic agent, X-glucuronide, which detects glucuronidase activity, an enzyme shown to be highly specific for Escherichia coli6.

Use TBX Medium for selective detection and enumeration of Escherichia coli in food and animal feed samples.

  • Selective: presence of bile salts in the medium provide selectivity against Gram-positive bacteria, particularly bacilli and fecal streptococci.
  • Easy-to-read: due to the addition of chromogenic 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide (X-glucuronide), E. coli appear as blue/green colonies.
  • ISO compliant: meets the formulation and performance criteria laid down in ISO 16649 and ISO 11133:2014.

Most Escherichia coli strains can be differentiated from other coliforms by the presence of the enzyme glucuronidase. However, approximately 3 to 4% of Escherichia coli are glucuronidase negative, notably Escherichia coli O157 strains7.

The chromogen in TBX Medium is 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide (X-glucuronide) and is targeted by glucuronidase. Escherichia coli cells are able to absorb this complex intact and intracellular glucuronidase splits the bond between the chromophore and the glucuronide. The released chromophore is colored and builds up within the cells, causing Escherichia coli colonies to be colored blue/green.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Especificaciones

Formulario Powder
Tipo de producto Dehydrated Culture Media
Cantidad 5 kg
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.