Learn More
Invitrogen™ TBP Monoclonal Antibody (1TBP18)
GREENER_CHOICE
Código de producto. 15682195
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
15682195 100 μL 100 microlitros
1 options
Código de producto. 15682195 Proveedor Invitrogen™ N.º de proveedor MA1189
Mouse Monoclonal Antibody
MA1-189 detects TATA-Binding Protein from human, mouse, and rat, canine, and non-human primate samples. TBP (human) has an approximate molecular weight of 38kDa, while other species' isoforms can range from 35-36kDa. MA1-189 has been successfully used in Western blot and immunofluorescence and flow cytometry applications. The MA1-189 immunogen is the N-terminal domain of the human TATA-binding protein (TBP).
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TBP
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 1TBP18
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4
génica Tbp
N.º de referencia del gen P20226, P29037
Alias de gen Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100)
Símbolos de los genes Tbp
Especie del huésped Mouse
Inmunógeno N-terminal domain of the human TATA-binding protein (TBP).
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117526, 21374, 611193, 6908
Especies diana Human, Mouse, Rat, Canine, Monkey
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1