missing translation for 'onlineSavingsMsg'
Learn More

TBK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Código de producto. 16856125 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16856125 150 μL 150 microlitros
16866125 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. View return policy
Product Code. 16856125 missing translation for 'mfr' Proteintech missing translation for 'supplierNo' 283971AP150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TBK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TBK1. This antibody recognizes human antigen. The TBK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antígeno TBK1
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Concentración 433 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 29110
Especies diana Human
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.