missing translation for 'onlineSavingsMsg'
Learn More

TBCC Monoclonal Antibody (OTI2B3), TrueMAB™, OriGene

Product Code. 17582242
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17582242 30 μL 30 microlitros
17562242 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17582242

Brand: Origene Technologies TA504709S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TBCC Monoclonal antibody specifically detects TBCC in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antígeno TBCC
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon OTI2B3
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica CPO
N.º de referencia del gen Q8IVL8
Alias de gen 2810055C19Rik; CFC; EGK_14912; Tbcc; tubulin folding cofactor C; tubulin-folding cofactor C; Tubulin-specific chaperone C
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human CPO produced in HEK293T cell.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 130749
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.