missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCC Monoclonal Antibody (OTI2B3), TrueMAB™, OriGene
Descripción
TBCC Monoclonal antibody specifically detects TBCC in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Especificaciones
Especificaciones
| Antígeno | TBCC |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2B3 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | CPO |
| N.º de referencia del gen | Q8IVL8 |
| Alias de gen | 2810055C19Rik; CFC; EGK_14912; Tbcc; tubulin folding cofactor C; tubulin-folding cofactor C; Tubulin-specific chaperone C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?