missing translation for 'onlineSavingsMsg'
Learn More

TBB, MedChemExpress

Código de producto. 16482187
Encompass_Preferred
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16482187

Marca: Medchem Express HY14394

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress TBB is a cell-permeable and ATP-competitive CK2 inhibitor with an IC50 of 0.15 μM for rat liver CK2.

TBB is a cell-permeable and ATP-competitive CK2 inhibitor with an IC50 of 0.15 μM for rat liver CK2.

Specifications

Nombre del producto químico o material TBB
CAS 17374-26-4
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C6HBr4N3
Cantidad 50 mg
Sinónimo NSC 231634 Casein Kinase II Inhibitor I
Información de solubilidad DMSO : ≥ 100 mg/mL (230.04 mM)
SMILES BrC1=C2C(NN=N2)=C(Br)C(Br)=C1Br
Peso molecular (g/mol) 434.71
Formula Weight (peso de la fórmula) 434.71
Porcentaje de pureza 99.31%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.