missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBATA Polyclonal Antibody

Descripción
This target displays homology in the following species: Human: 100%.
This gene encodes a protein that regulates thymic epithelial cell proliferation and thymus size. It has been identified as a ligand for the class I human leukocyte antigen (HLA-I) in thymus. Studies of the orthologous mouse protein suggest that it may also play a role in spermatid differentiation, as well as in neuronal morphogenesis and synaptic plasticity. Polymorphisms in this gene are associated with susceptibility for multiple sclerosis (MS). Alternative splicing results in multiple transcript variants.
Especificaciones
Especificaciones
| Antígeno | TBATA |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | TBATA |
| N.º de referencia del gen | Q96M53 |
| Alias de gen | 1700021K02Rik; AI428928; C10orf27; protein SPATIAL; Protein TBATA; RGD1561665; SPATIAL; stromal protein associated with thymii and lymph node; Stromal protein associated with thymii and lymph node homolog; stromal protein associated with thymii and lymph nodes; TBATA; thymus and testis specific nuclear protein; thymus, brain and testes associated; thymus, brain and testes-associated protein; Titest |
| Símbolos de los genes | TBATA |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?