Descripción
May regulate a number of protein-protein interactions by competing for PDZ domain binding sites. Binds CTNNB1 and may thereby act as an inhibitor of the Wnt signaling pathway. Competes with LIN7A for KCNJ4 binding, and thereby promotes KCNJ4 internalization. May play a role in the Rho signaling pathway. May play a role in activation of CDC42 by the viral protein HPV16 E6.
Especificaciones
Especificaciones
| Antígeno | TAX1BP3 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.47 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | TAX1BP3 |
| N.º de referencia del gen | O14907, Q4QQV1, Q9DBG9 |
| Alias de gen | Tax interaction protein 1, Tax1 binding protein 3, TAX1BP3, TIP 1, TIP1 |
| Símbolos de los genes | TAX1BP3 |
| Mostrar más |