missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA578594
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positive Control: K562, K562 MF Predicted Reactivity: Pig (80%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by T1R3 is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.
Especificaciones
| TAS1R3 | |
| Polyclonal | |
| Unconjugated | |
| TAS1R3 | |
| Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 0.1M glycine, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7RTX0 | |
| TAS1R3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human T1R3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido