Learn More
Invitrogen™ Talquetamab Humanized Recombinant Human Monoclonal Antibody
Descripción
Talquetamab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Talquetamab (JNJ-64407564) is a bispecific antibody that targets CD3, which is expressed on T-cells, and GPRC5D, which is highly expressed in multiple myeloma cells. Talquetamab (JNJ-64407564) is being investigated in a Phase 1 clinical study to treat relapsed or refractory multiple myeloma.
Especificaciones
Especificaciones
| Antígeno | Talquetamab Humanized |
| Aplicaciones | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Clasificación | Recombinant Monoclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Alias de gen | JNJ-64407564 |
| Especie del huésped | Human |
| Método de purificación | Protein A |
| Cantidad | 1 mg |
| Mostrar más |