missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF4 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17980911
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
17980911 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17980911

Marca: Invitrogen™ PA5143943

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human A431 whole cell, human K562 whole cell, human Caco-2 whole cell, human U20S whole cell, human HepG2 whole cell, human PC-3 whole cell, rat brain tissue, mouse brain tissue. IHC: human ovarian serous adenocarcinoma tissue, human ovarian cancer tissue, human breast cancer tissue, human gastric adenocarcinoma tissue, human renal clear cell carcinoma tissue, human tonsil tissue, mouse brain tissue, rat brain tissue, human gastric adenocarcinoma tissue, human breast cancer tissue. ICC/IF: CACO-2 cell. Flow: Hela cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TAF4
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 500 μg/mL
Conjugado Unconjugated
Formulación PBS with 4mg trehalose and no preservative
génica TAF4
N.º de referencia del gen O00268
Alias de gen 135kDa; AI450312; FLJ41943; LOW QUALITY PROTEIN: transcription initiation factor TFIID subunit 4; RNA polymerase II TBP-associated factor subunit C; TAF(II)130; TAF(II)135; TAF2C; TAF2C1; TAF4; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135kDa; Taf4a; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135 kD; TAFII 130; TAFII 135; TAFII130; TAFII-130; TAFII135; TAFII-135; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C1, 130kD; TATA box binding protein associated factor 4; TATA-box binding protein associated factor 4; TBP-associated factor 4; Transcription initiation factor TFIID 130 kDa subunit; transcription initiation factor TFIID 135 kD subunit; transcription initiation factor TFIID 135 kDa subunit; Transcription initiation factor TFIID subunit 4
Símbolos de los genes TAF4
Especie del huésped Rabbit
Inmunógeno E.coli-derived human TAF4 recombinant protein (Position: D6-K1085).
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 228980, 682097, 6874
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.