Learn More
TAF1B Mouse anti-Human, Clone: OTI2A12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF808691
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of RNA polymerase I core factor complex that acts as a GTF2B/TFIIB-like factor and plays a key role in multiple steps during trancription initiation such as preinitiation complex (PIC) assembly and postpolymerase recruitment events in polymerase I (Pol I) transcription. Binds rDNA promoters and plays a role in Pol I recruitment as a component of the SL1/TIF-IB complex and, possibly, directly through its interaction with RRN3.Especificaciones
| TAF1B | |
| Monoclonal | |
| Unconjugated | |
| TAF1B | |
| MGC:9349, RAF1B, RAFI63, SL1, TAFI63 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9014 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2A12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q53T94 | |
| TAF1B | |
| Human recombit protein fragment corresponding to amino acids 1-208 of human TAF1B produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.