missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14410-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TAB2 is an activator of MAP3K7/TAK1, which is required for for the IL-1 induced activation NF-kappaB and MAPK8/JNK. This protein forms a kinase complex with TRAF6, MAP3K7 and TAB1, thus serves as an adaptor linking MAP3K7 and TRAF6. This protein, TAB1, and MAP3K7 also participate in the signal transduction induced by TNFSF11/RANKL through the activation of the receptor activator of NF-kappaB (TNFRSF11A/RANK), which may regulate the development and function of osteoclasts. Recent experiments have shown that TAB2 and the related protein TAB3 constitutitvely interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.Especificaciones
| TAB2 | |
| Polyclonal | |
| Unconjugated | |
| TAB2 | |
| KIAA0733, MAP3K7IP2, TAB 2, TAB2, TAK1 binding protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23118, 308267, 68652 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U303, Q99K90, Q9NYJ8 | |
| Tab2 | |
| TAB2 Fusion Protein Ag5742 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido