Learn More
Thermo Scientific™ T4 DNA Ligase PROMO
T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.
Marca:  Thermo Scientific™ EL0016-RMD123
Código de producto. 15699444
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
Highlights
  • Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
  • Fast – sticky-end ligation is completed in 10 minutes at room temperature
  • Supplied with PEG solution for efficient blunt-end ligation
Applications
  • Cloning of restriction enzyme generated DNA fragments
  • Cloning of PCR products
  • Joining of double-stranded oligonucleotide linkers or adaptors to DNA
  • Site-directed mutagenesis
  • Amplified fragment length polymorphism (AFLP)
  • Ligase-mediated RNA detection (see Reference 3)
  • Nick repair in duplex DNA, RNA or DNA/RNA hybrids
  • Self-circularization of linear DNA.
Notes
  • Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
  • The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
  • Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture by spin column or chloroform extraction. The extracted DNA can be further precipitated with ethanol.