missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, APC, Clone: GL3, STEMCELL Technologies
Descripción
- The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
- TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
- It is a member of the immunoglobulin superfamily
- TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
- These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
- Once activated, they secrete effector cytokines in a subtype- and context-specific manner
- Most γ/δ T cells are CD4-/CD8-, though some express CD8
- The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Especificaciones
Especificaciones
| Antígeno | T Cell Receptor Gamma/Delta |
| Aplicaciones | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay |
| Clasificación | Monoclonal |
| Clon | GL3 |
| Conjugado | APC |
| Especie del huésped | Hamster |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Isotype | IgG2 κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?