Learn More
CELLPATH SYSTEM III BIOPSY CASSETTE
Código de producto. p-200113600
Change view
Click to view available options
Material:
Loose
Threaded
Tamaño de la unidad:
paquete de 2000
paquete de 500
Código de producto. Material unitSize
30226236 Loose paquete de 500
30226237 Threaded paquete de 2000
2 options
Código de producto. 30226236
Marca: CELLPATH EAI060210A
The System III Biopsy Cassette comes with an attached lid with an easy to break-off hinge. Incorporating square apertures of 0.9 mm by 0.9 mm, the Biopsy Cassette ensures safe accommodation of smaller biopsy specimens, whilst also enabling good levels of fluid transfer.
  • The locking mechanism of the cassette ensures maximum tissue security during processing.
  • The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
  • Key Benefits:
  • Compatible with Primera, Leica and Sakura Printers
  • Optimised writing surface
  • Increased reagent flow
  • Integral fixed frangible lid
  • High rigidity and stability
Especificaciones
Color White
Material Loose
Tipo de producto Cassette