Learn More
CELLPATH SYSTEM II+ SLOTTED CASSETTE
Código de producto. p-200113602
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Tamaño de la unidad:
paquete de 1000
paquete de 2000
paquete de 3000
Código de producto. Material unitSize
30226240 Loose paquete de 1000
30226241 Hoppered paquete de 3000
30226242 Threaded paquete de 2000
3 options
Código de producto. 30226240
Marca: CELLPATH EAM050203A
Edge
Incorporating 1.1 mm wide slots, the System II+ Slotted Cassette increases reagent flow, facilitating improved consistency and quality of processed specimens. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Especificaciones
Color White
Material Loose
Tipo de producto Cassette