Learn More
CELLPATH SYSTEM II+ BIOPSY CASSETTE
Código de producto. p-200113603
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Tamaño de la unidad:
paquete de 1000
paquete de 2000
paquete de 3000
Código de producto. Material unitSize
30226243 Loose paquete de 1000
30226244 Hoppered paquete de 3000
30226245 Threaded paquete de 2000
3 options
Código de producto. 30226243
Marca: CELLPATH EAM060203A
Incorporating 0.9 x 0.9 mm apertures, the System II+ Biopsy Cassette offers improved specimen security for small biopsy and tissue fragments. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Especificaciones
Color White
Material Loose
Tipo de producto Cassette