missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin II Recombinant Rabbit Monoclonal Antibody (23GB4960), Invitrogen™

Description
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
Specifications
| Antígeno | Synapsin II |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB4960 |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | SYN2 |
| N.º de referencia del gen | Q63537, Q92777 |
| Alias de gen | 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb |
| Símbolos de los genes | SYN2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?