missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syk Polyclonal Antibody

Descripción
Recommended positive controls: A431, Raji, mouse BMDM, mouse spleen. Predicted reactivity: Mouse (87%), Rat (87%), Pig (89%), Rhesus Monkey (98%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.
Especificaciones
Especificaciones
| Antígeno | Syk |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.32 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| génica | Syk |
| N.º de referencia del gen | P43405, P48025, Q64725 |
| Alias de gen | DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK |
| Símbolos de los genes | Syk |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?