missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420H2 Antibody (RAB-C409) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals NBP3-20150-0.025mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SUV420H2 Monoclonal antibody specifically detects SUV420H2 in Human samples. It is validated for ELISA, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Especificaciones
| SUV420H2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Human | |
| Affinity purified | |
| RUO | |
| 84787 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| ELISA, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| RAB-C409 | |
| Unconjugated | |
| EC 2.1.1.43, FLJ98627, histone-lysine N-methyltransferase SUV420H2, KMT5Csuv4-20h2, Lysine N-methyltransferase 5C, MGC2705, Su(var)4-20 homolog 2, Suppressor of variegation 4-20 homolog 2, suppressor of variegation 4-20 homolog 2 (Drosophila), Suv4-20h2 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SUV420H2 protein under non-denaturing conditions. | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido