missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard: S @ 50000 μg/g (5.00 wt%) in Crude Oil, VHG™

Código de producto. 30040899
Change view
Click to view available options
Cantidad:
50 mL
100 mL
500 mL
Tamaño de la unidad:
100 ml
50 ml
500 ml
Código de producto. Cantidad unitSize
30040899 500 mL 500 ml
30040643 100 mL 100 ml
30040512 50 mL 50 ml
3 options
Este artículo no se puede devolver. View return policy

Product Code. 30040899

Brand: VHG Labs VHGSPCRD5P500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur Standard: S @ 50000 μg/g (5.00 wt%) in Crude Oil
TRUSTED_SUSTAINABILITY

Specifications

Nombre del producto químico o material Sulfur
CAS 7704-34-9
Métodos EDXRF/WDXRF
Nombres de analitos o componentes Sulfur
Condiciones de envío Room Temperature
Fórmula molecular S
Concentración 5%
Certificaciones/conformidad ISO 17034
Fórmula InChI InChI=1S/S
SMILES [S]
Nombre IUPAC sulfur
Solvent or Matrix Crude Oil
Peso molecular (g/mol) 32.065
Cantidad 500 mL
Formula Weight (peso de la fórmula) 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.