missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sugemalimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Código de producto. 30283612
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
Código de producto. Cantidad unitSize
30283612 100 μg 100 microgramos
30283605 1 mg 1 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30283612

Marca: Invitrogen™ MA559121

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Sugemalimab, also known as Cejemly, is a fully human, full-length IgG4 monoclonal antibody targeting programmed death-ligand 1 (PD-L1). The gene PD-L1 (Programmed Death-Ligand 1), also known as CD274, encodes a protein crucial for regulating immune responses. Located on chromosome 9, PD-L1 is expressed on the surface of various cells, including tumor cells, immune cells, and other tissues. PD-L1 functions by binding to its receptor PD-1 on T cells, leading to the inhibition of T cell activity and induction of immune tolerance. This interaction plays a pivotal role in preventing autoimmunity and promoting immune escape in cancer, allowing tumor cells to evade the immune system. The expression of PD-L1 is upregulated in many cancers, including melanoma, lung cancer, and renal cell carcinoma, making it a critical target for cancer immunotherapies. Monoclonal antibodies targeting PD-L1, such as atezolizumab and durvalumab, have shown efficacy in restoring anti-tumor immunity. Further research into the regulation and function of PD-L1 continues to enhance our understanding of its role in immune modulation and its potential as a therapeutic target.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Sugemalimab
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.