Learn More
Solución estándar Brix de sacarosa, 20 %, Reagecon™
BIOPHARMA
Product Code. 13494046 Shop All Reagecon Products
Change view
Click to view available options
Cantidad:
15 mL
Unit Size:
paquete de 6
Product Code. Cantidad unitSize
13494046 15 mL paquete de 6
1 options
This item is not returnable. View return policy
Product Code. 13494046
Brand: Reagecon BS20S6